missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Somatostatin R5/SSTR5 Alexa Fluor 350-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC4448U-100UG
This item is not returnable.
View return policy
Description
Somatostatin R5/SSTR5 Monoclonal specifically detects Somatostatin R5/SSTR5 in Human samples. It is validated for Flow Cytometry.
Specifications
| Somatostatin R5/SSTR5 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| AAK61266 | |
| SSTR5 | |
| NS0 mouse myeloma cell line transfected with human Somatostatin R5/SSTR5 Met1-Leu364 Accession # AAK61266 | |
| 100 μg | |
| Primary | |
| Detects human Somatostatin R5/SSTR5 in direct ELISAs. Stains Somatostatin R5/SSTR5 transfectants but not irrelevant transfectants. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG1 |
| Flow Cytometry | |
| 394401 | |
| Alexa Fluor 350 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| somatostatin receptor 5, somatostatin receptor subtype 5, somatostatin receptor type 5, SomatostatinR5, SS5R, SS-5-R, SS5-R, SST5, SSTR5 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6755 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction