missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Somatostatin Antibody, R&D Systems™
Rabbit Monoclonal Antibody
£206.00 - £467.00
Specifications
| Antigen | Somatostatin |
|---|---|
| Clone | 2957D |
| Dilution | Immunohistochemistry 1-10 μg/mL |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30279874
|
R&D Systems
MAB11635-SP |
25 μg |
£206.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30279865
|
R&D Systems
MAB11635-100 |
100 μg |
£467.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Somatostatin Monoclonal antibody specifically detects Somatostatin in Human samples. It is validated for ImmunohistochemistrySpecifications
| Somatostatin | |
| Immunohistochemistry 1-10 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. | |
| Growth hormone release-inhibiting factor, SMST, SOM, somatostatin, somatostatin-14, somatostatin-28, SRIF, SST | |
| Synthetic Peptide, Accession # P61278 | |
| Primary | |
| Reconstitute lyophilized material at 0.2 mg/mL in sterile PBS. For liquid material, refer to CoA for concentration. |
| 2957D | |
| Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P61278 | |
| 6750 | |
| IgG | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20°C to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title