missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Human Soluble TWEAK/TNFSF12 ELISA Kit (Colorimetric)

Product Code. 18237161 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18237161 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18237161 Supplier Novus Biologicals™ Supplier No. NBP2601451KIT

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High sensitivity and specificity for detection of human sTWEAK.

The Human Soluble TWEAK/TNFSF12 ELISA Kit (Colorimetric) measures human Soluble TWEAK/TNFSF12 in serum, plasma, tissue homogenates.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 31.25 to 2000pg/mL
Assay Sensitivity 7.8pg/mL
Product Type ELISA Kit (Colorimetric)
Sample Type Plasma, Serum, Tissue Homogenates
Specificity For detection of human sTWEAK
Target Species Human
For Use With (Application) ELISA
Kit Contents Adhesive Strip (For 96 wells), Assay plate (12 x 8 coated Microwells), Biotin-antibody (100 x concentrate), Biotin-antibody Diluent, HRP-avidin (100 x concentrate), HRP-avidin Diluent, TMB Substrate, Wash Buffer (25 x concentrate), Stop Solution, Standard (Freeze dried)Sample Diluent, Instruction manual
Quantity 1 Kit
Regulatory Status RUO
Research Discipline Cardiovascular Biology
Sample Volume 50 to 100 μL
Storage Requirements Store the unopened product at 2° to 8°C. Protect from light. Do not use past expiration date.
Target Soluble TWEAK/TNFSF12
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.