missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human SOD3/EC-SOD Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB34201-SP
This item is not returnable.
View return policy
Description
SOD3/EC-SOD Monoclonal specifically detects SOD3/EC-SOD in Human samples. It is validated for Western Blot.
Specifications
| SOD3/EC-SOD | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| P08294 | |
| SOD3 | |
| E. coli-derived recombinant human SOD3/EC-SOD Trp19-Ala240 Accession # P08294 | |
| 25 μg | |
| Primary | |
| Detects human SOD3/EC-SOD in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human SOD2, recombinant mouse (rm) SOD1, or rmSOD3 is observed. | |
| Human | |
| Purified |
| Western Blot | |
| 713707 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ECSOD, EC-SOD, EC-SODEC 1.15.1.1, extracellular superoxide dismutase [Cu-Zn], MGC20077, superoxide dismutase 3, extracellular | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6649 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction