missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human SOCS-7/Nck/NAP4 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB5809-SP
This item is not returnable.
View return policy
Description
SOCS-7/Nck/NAP4 Monoclonal specifically detects SOCS-7/Nck/NAP4 in Human samples. It is validated for Western Blot.
Specifications
| SOCS-7/Nck/NAP4 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.5 ug/mL | |
| O14512 | |
| SOCS7 | |
| E. coli-derived recombinant human SOCS-7 His351-Thr581 Accession # O14512 | |
| 25 μg | |
| Primary | |
| Detects endogenous human SOCS-7 in Western blots. | |
| Human | |
| Purified |
| Western Blot | |
| 519106 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| NAP4, NAP-4, NAP4SOCS-7, Nck, Nck, Ash and phospholipase C binding protein, Nck, Ash and phospholipase C gamma-binding protein, NCKAP4, Nck-associated protein 4, SOCS6, SOCS7, suppressor of cytokine signaling 7 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 30837 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction