missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human SNRPA (aa 173-216) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
The protein encoded by this gene associates with stem loop II of the U1 small nuclear ribonucleoprotein, which binds the 5' splice site of precursor mRNAs and is required for splicing. The encoded protein autoregulates itself by polyadenylation inhibition of its own pre-mRNA via dimerization and has been implicated in the coupling of splicing and polyadenylation.
Specifications
Specifications
| Accession Number | P09012 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 6626 |
| Name | Human SNRPA (aa 173-216) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | C430021M15Rik; fc19d01; MGC137110 protein; Mud1; nRNP A; nuclear ribonucleoprotein A; protein A of the U1 small nuclear RNA; Rnu1a1; Rnu1a-1; small nuclear ribonucleoprotein polypeptide A; Snrpa; U1 small nuclear ribonucleoprotein A; U1 small nuclear RNP-specific A; U1 snRNP A; U1 snRNP protein A; U1 snRNP-specific protein A; U1A; U1-A; Unknown (protein for MGC:77810); wu:fc19d01; zgc:77810 |
| Common Name | SNRPA |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction