missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human SNAP29 (aa 186-248) Control Fragment Recombinant Protein

Product Code. 30201681
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30201681

Brand: Invitrogen™ RP95340

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (89%), Rat (89%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-56870 (PA5-56870. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

SNAP29 encodes a product that is a member of the SNAP25 gene family, encodes a protein involved in multiple membrane trafficking steps. Two other members of this gene family, SNAP23 and SNAP25, encode proteins that bind a syntaxin protein and mediate synaptic vesicle membrane docking and fusion to the plasma membrane. The protein encoded by this gene binds tightly to multiple syntaxins and is localized to intracellular membrane structures rather than to the plasma membrane. While the protein is mostly membrane-bound, a significant fraction of it is found free in the cytoplasm. Use of multiple polyadenylation sites has been noted for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O95721
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 9342
Name Human SNAP29 (aa 186-248) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 1300018G05Rik; AI891940; AU020222; BB131856; CEDNIK; fc84f09; FLJ21051; golgi SNARE of 32 kDa; Gs32; im:7145101; LOC553460 protein; si:dkey-178e17.4; SNAP29; SNAP-29; Soluble 29 kDa NSF attachment protein; synaptosomal-associated protein 29; synaptosomal-associated protein 29 kD; synaptosomal-associated protein, 29 kD; synaptosomal-associated protein, 29 kDa; synaptosome associated protein 29; synaptosome associated protein 29 kDa; vesicle-membrane fusion protein SNAP-29; wu:fc84f09
Common Name SNAP29
Gene Symbol Snap29
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence TDAYPKNPHLRAYHQKIDSNLDELSMGLGRLKDIALGMQTEIEEQDDILDRLTTKVDKLDVNI
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.