missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human SNAP25 Antibody, R&D Systems™
Mouse Monoclonal Antibody
£222.00 - £491.00
Specifications
| Antigen | SNAP25 |
|---|---|
| Clone | 1065704 |
| Dilution | Immunocytochemistry 8-25 μg/mL |
| Applications | Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18774404
|
R&D Systems
MAB11418-100 |
100 μg |
£491.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18774474
|
R&D Systems
MAB11418-SP |
25 μg |
£222.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SNAP25 Monoclonal antibody specifically detects SNAP25 in Human samples. It is validated for ImmunocytochemistrySpecifications
| SNAP25 | |
| Immunocytochemistry 8-25 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| bA416N4.2, dJ1068F16.2, FLJ23079, resistance to inhibitors of cholinesterase 4 homolog, RIC4, RIC-4, SEC9, SNAP-25SNAP, SUP, Super protein, Synaptosomal-associated 25 kDa protein, synaptosomal-associated protein 25, synaptosomal-associated protein, 25kD, synaptosomal-associated protein, 25kDa | |
| E. coli -derived recombinant human SNAP-25, Asp140-Gly206, Accession # P60880 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 1065704 | |
| Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| P60880 | |
| 6616 | |
| IgG3 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title