missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Snail Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 9 publications
Brand: R&D Systems AF3639-SP
This item is not returnable.
View return policy
Description
Snail Polyclonal specifically detects Snail in Human samples. It is validated for Western Blot, Intracellular Staining by Flow Cytometry, Chromatin Immunoprecipitation (ChIP), Immunocytochemistry.
Specifications
| Snail | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Protein sna, Protein snail homolog 1, SLUGH2, SNA, SNAH, SNAHdJ710H13.1, SNAI1, snail 1 (drosophila homolog), zinc finger protein, snail 1 homolog, snail 1 zinc finger protein, snail 1, zinc finger protein, snail homolog 1 (Drosophila), zinc finger protein SNAI1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 6615 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| ChIP Assay, Flow Cytometry, Immunocytochemistry | |
| LYOPH | |
| Western Blot 0.5 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, Chromatin Immunoprecipitation (ChIP) 5 ug/5 x 10^6 cells, Immunocytochemistry 5-15 ug/mL | |
| O95863 | |
| SNAI1 | |
| E. coli-derived recombinant human Snail Pro2-Arg264 Accession # O95863 | |
| 25 μg | |
| Primary | |
| Detects human Snail in direct ELISAs and Western blots. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction