missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human SNAIL (aa 117-167) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (82%), Rat (82%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Zinc finger protein SNAI1 (SNAIL)is a zinc finger transcriptional repressor which down regulates the expression of the ectodermal gene within the mesoderm and functions to form the mesoderm and neural crest. It is reported that SNAI1 acts as an important factor of tumor invasion as evidenced by its role in E-cadherin down-regulation and induction of epithelial-mesenchymal transition (EMT). In human breast cancer, the expression of SNAI1 and/or the homologous SNAI2 (Slug) has been associated with E-cadherin repression, local or distant metastasis, tumor recurrence, or poor prognosis in different tumor series. It is also reported that Snail 1 protein in the stromamay be anew putative prognosis marker for colon tumors.
Specifications
Specifications
| Accession Number | O95863 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 6615 |
| Name | Human SNAIL (aa 117-167) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | AI194338; dJ710H13.1; Protein sna; protein snail homolog 1; SLUGH2; SNA; Sna protein; Sna1; SNAH; snai; SNAI1; Snail; snail 1; snail 1 homolog; snail 1 zinc finger protein; snail 1, zinc finger protein; snail family transcriptional repressor 1; snail family zinc finger 1; snail homolog 1; Snail1; Zinc finger protein SNAI1 |
| Common Name | SNAIL |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction