missing translation for 'onlineSavingsMsg'
Learn More

Human Smoothened Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody

£168.00 - £416.00

Specifications

Antigen Smoothened
Clone 1015418
Dilution Immunohistochemistry 1-25 ug/mL
Applications Immunohistochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18760877
GREENER_CHOICE
View Documents
R&D Systems
MAB10276-SP
25 μg
£168.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18752307
GREENER_CHOICE
View Documents
R&D Systems
MAB10276-100
100 μg
£416.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Smoothened Monoclonal specifically detects Smoothened in Human samples. It is validated for Immunohistochemistry.
Specifications

Specifications

Smoothened
Immunohistochemistry 1-25 ug/mL
Monoclonal
Purified
RUO
Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
bnb, CRJS, FZD11, Gx, Protein Gx, SMO, SMOH, SMOHseven transmembrane helix receptor, smoothened (Drosophila) homolog, smoothened homolog, smoothened homolog (Drosophila)
Chinese Hamster Ovary cell line, CHO-derived human Smoothened, Met1-Tyr233, Accession # Q99835
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human Smoothened in direct ELISAs.
1015418
Immunohistochemistry
Unconjugated
Mouse
Human
Q99835
6608
IgG2a
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.