missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human SMNDC1 (aa 165-220) Control Fragment Recombinant Protein

Product Code. 30212252
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30212252

Brand: Invitrogen™ RP109217

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene is a paralog of SMN1 gene, which encodes the survival motor neuron protein, mutations in which are cause of autosomal recessive proximal spinal muscular atrophy. The protein encoded by this gene is a nuclear protein that has been identified as a constituent of the spliceosome complex. This gene is differentially expressed, with abundant levels in skeletal muscle, and may share similar cellular function as the SMN1 gene.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O75940
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 10285
Name Human SMNDC1 (aa 165-220) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 2410004J23Rik; 30 kDa splicing factor SMNrp; 4933440I19Rik; MGC106917; MGC112663; SMN related protein; SMNDC1; SMNR; SMN-related protein; Spf30; splicing factor 30, survival of motor neuron-related; survival motor neuron domain containing 1; survival motor neuron domain-containing protein 1; Survival of motor neuron-related-splicing factor 30; TDRD10; TDRD16C; tudor domain containing 16 C; wu:fb37h07; wu:fc23a07
Common Name SMNDC1
Gene Symbol SMNDC1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence KELEQEREDQKVKWQQFNNRAYSKNKKGQVKRSIFASPESVTGKVGVGTCGIADKP
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.