missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human SLX4 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
£185.00 - £432.00
Specifications
| Antigen | SLX4 |
|---|---|
| Clone | 2392A |
| Dilution | Immunocytochemistry 3-25 ug/mL |
| Applications | Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18742467
![]() |
R&D Systems
MAB11246-SP |
25 μg |
£185.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18770217
![]() |
R&D Systems
MAB11246-100 |
100 μg |
£432.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SLX4 Monoclonal antibody specifically detects SLX4 in Human samples. It is validated for ImmunocytochemistrySpecifications
| SLX4 | |
| Immunocytochemistry 3-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| Synthetic peptide within amino acids 400-500 of the Human SLX4 protein sequence, | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 2392A | |
| Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| SLX4 | |
| IgG | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title