missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human SLURP1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB4479
This item is not returnable.
View return policy
Description
SLURP1 Monoclonal specifically detects SLURP1 in Human samples. It is validated for Immunohistochemistry.
Specifications
| SLURP1 | |
| Monoclonal | |
| LYOPH | |
| Immunohistochemistry 8-25 ug/mL | |
| P55000 | |
| SLURP1 | |
| E. coli-derived recombinant human SLURP1 Ala22-Leu103 Accession # P55000 | |
| 100 μg | |
| Primary | |
| Detects human SLURP1 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse SLURP1 is observed. | |
| Human | |
| Purified |
| Immunohistochemistry | |
| 569317 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ANUPARS(component B)-81/S, ARS component B, ArsB, ARSlymphocyte antigen 6-like secreted, LY6LS, MDMsecreted Ly-6/uPAR-related protein 1, neoplastic urinary protein, secreted LY6/PLAUR domain containing 1, secreted Ly6/uPAR related protein 1, SLURP-1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 57152 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction