missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human SLPI Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB1274
This item is not returnable.
View return policy
Description
SLPI Monoclonal specifically detects SLPI in Human samples. It is validated for Western Blot.
Specifications
| SLPI | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| P03973 | |
| SLPI | |
| E. coli-derived recombinant human SLPI Accession # P03973 | |
| 500 μg | |
| Primary | |
| Detects recombinant human SLPI in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant mouse SLPI is observed. | |
| Human | |
| Purified |
| Western Blot | |
| 20409 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ALK1, ALPProtease inhibitor WAP4, antileukoproteinase, BLPISeminal proteinase inhibitor, HUSI, HUSI-I, Mucus proteinase inhibitor, secretory leukocyte peptidase inhibitor, Secretory leukocyte protease inhibitor, secretory leukocyte protease inhibitor (antileukoproteinase), WAP four-disulfide core domain protein 4, WAP4HUSI-1, WFDC4MPI | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6590 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction