Learn More
Human SLITRK5 Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand:  R&D Systems AF2587
Additional Details : Weight : 0.00970kg
 View more versions of this product
Product Code. 18001312
  • £428.28 / 100µg
Estimated Shipment: 03-06-2024
to see stock.
Description
Description
SLITRK5 Polyclonal specifically detects SLITRK5 in Human samples. It is validated for Western Blot.
Specifications
Specifications
SLITRK5
Polyclonal
Unconjugated
O94991
SLITRK5
Mouse myeloma cell line NS0-derived recombinant human SLITRK5 Glu39-Val665 Accession # O94991
100 μg
Primary
Detects human SLITRK5 in direct ELISAs and Western blots. In direct ELISAs, less than 2% cross-reactivity with recombinant human (rh) SLITRK1, rhSLITRK2, rhSLITRK3, rhSLITRK4 and rhSLITRK6 is observed.
Human
IgG
Western Blot
LYOPH
Western Blot 0.1 ug/mL
bA364G4.2, FLJ58374, KIAA0918, leucine-rich repeat-containing protein 11, LRRC11, SLIT and NTRK-like family, member 5, SLIT and NTRK-like protein 5
Goat
Affinity Purified
RUO
26050
Reconstitute at 0.2 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.