missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human SLITRK5 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB2587
Additional Details : Weight : 0.00970kg
Description
SLITRK5 Monoclonal specifically detects SLITRK5 in Human samples. It is validated for Western Blot.Specifications
SLITRK5 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
O94991 | |
SLITRK5 | |
Mouse myeloma cell line NS0-derived recombinant human SLITRK5 Glu39-Val665 Accession # O94991 | |
100 μg | |
Primary | |
Detects human SLITRK5 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human SLITRK1, 2, 3, or 4 is observed. | |
Human | |
Purified |
Western Blot | |
310516 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
bA364G4.2, FLJ58374, KIAA0918, leucine-rich repeat-containing protein 11, LRRC11, SLIT and NTRK-like family, member 5, SLIT and NTRK-like protein 5 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
26050 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
For Research Use Only