missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human SLITRK1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB3009
This item is not returnable.
View return policy
Description
SLITRK1 Monoclonal specifically detects SLITRK1 in Human samples. It is validated for Western Blot.
Specifications
| SLITRK1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q96PX8 | |
| SLITRK1 | |
| Mouse myeloma cell line NS0-derived recombinant human SLITRK1 Ala16-Ser616 Accession # Q96PX8 | |
| 100 μg | |
| Primary | |
| Detects human SLITRK1 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) SLITRK2, rhSLITRK4, or rhSLITRK5 is observed. | |
| Human | |
| Purified |
| Western Blot | |
| 327808 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| FLJ54428, KIAA1910KIAA0918, leucine rich repeat containing 12, Leucine-rich repeat-containing protein 12, LRRC12, RP11-395N17.1, SLIT and NTRK-like family, member 1, SLIT and NTRK-like protein 1, slit and trk like gene 1, TTM | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 114798 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction