missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human SLITRK1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£157.00 - £337.00
Specifications
| Antigen | SLITRK1 |
|---|---|
| Clone | 327829 |
| Concentration | LYOPH |
| Dilution | Immunohistochemistry 8-25 ug/mL |
| Applications | Immunohistochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18797209
![]() |
R&D Systems
MAB30091-SP |
25 μg |
£157.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18131171
|
R&D Systems
MAB30091 |
100 μg |
£337.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SLITRK1 Monoclonal specifically detects SLITRK1 in Human samples. It is validated for Immunohistochemistry.Specifications
| SLITRK1 | |
| LYOPH | |
| Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q96PX8 | |
| 114798 | |
| Mouse myeloma cell line NS0-derived recombinant human SLITRK1 Ala16-Ser616 Accession # Q96PX8 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human SLITRK1 in direct ELISAs. In direct ELISAs, no cross-reactivity with reactivity with SLITRK2, 3, 4, or 5 is observed. |
| 327829 | |
| Immunohistochemistry 8-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| FLJ54428, KIAA1910KIAA0918, leucine rich repeat containing 12, Leucine-rich repeat-containing protein 12, LRRC12, RP11-395N17.1, SLIT and NTRK-like family, member 1, SLIT and NTRK-like protein 1, slit and trk like gene 1, TTM | |
| SLITRK1 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title