missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human SLITRK1 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 1 publication
Brand: R&D Systems AF3009
Additional Details : Weight : 0.00970kg
Description
SLITRK1 Polyclonal specifically detects SLITRK1 in Human samples. It is validated for Western Blot.Specifications
SLITRK1 | |
Polyclonal | |
Unconjugated | |
Q96PX8 | |
SLITRK1 | |
Mouse myeloma cell line NS0-derived recombinant human SLITRK1 (R&D Systems, Catalog # 3009-SK) Ala16-Ser616 Accession # Q96PX8 | |
100 μg | |
Primary | |
Detects human SLITRK1 in direct ELISAs and Western blots. In these formats, approximately 5% cross-reactivity with recombinant human (rh) SLITRK2, rhSLITRK4, and rhSLITRK5 is observed. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
FLJ54428, KIAA1910KIAA0918, leucine rich repeat containing 12, Leucine-rich repeat-containing protein 12, LRRC12, RP11-395N17.1, SLIT and NTRK-like family, member 1, SLIT and NTRK-like protein 1, slit and trk like gene 1, TTM | |
Goat | |
Affinity Purified | |
RUO | |
114798 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only