missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human Slit2-N Recombinant Protein, PeproTech®

Product Code. 17879902
Change view
Click to view available options
Quantity:
100 μg
2.5 x 200 μg
200 μg
25 μg
5 μg
5 x 200 μg
50 μg
Unit Size:
100µg
1mg
200µg
25µg
500µg
50µg
5µg
Pack of 5
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17879902 5 μg 5µg
17840923 100 μg 100µg
17860943 5 x 200 μg 1mg
30088198 5 x 200 μg Pack of 5
30087536 2.5 x 200 μg 500µg
17820753 200 μg 200µg
17840113 25 μg 25µg
17850923 2.5 x 200 μg 500µg
17830363 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Product Code. 17879902 Supplier Gibco™ Supplier No. 150115UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

150-11-1MG will be provided as 5 x 200 μg (150-11-200UG). 150-11-500UG will be provided as multiple 100 μg (150-11-100UG) and 200 μg (150-11-200UG) vials. Recombinant Human Slit2-N is a 1093 amino acid glycoprotein corresponding to the N-terminal portion of the full length Slit2 precursor, and has a calculated, theoretical molecular weight of 122.35 kDa. Due to glycosylation Slit2-N migrates at an apparent molecular weight of approximately 120.0-140.0 kDa by SDS-PAGE analysis under reducing conditions. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

SLIT2 are ligands for Robo receptors and play a vital role for axonal migration/navigation during neural development. A number of cleavage products through alternate splicing are reported in the literature for SLIT2 proteins. The C-terminal cleavage proteins are more diffusible than the larger N-terminal protein that is more tightly cell associated. SLIT2 protein is expressed in tissues such as fetal lung, kidney, and adult spinal cord.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O94813
For Use With (Application) Control, Bioactivity
Formulation Protein with no preservative
Gene ID (Entrez) 9353
Molecular Weight (g/mol) 122.35 kDa
Name Human Slit2-N
Purification Method Purified
Quantity 5 μg
Source HEK293 cells
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias downregulated during adipocyte differentiation-1; Drad-1; E030015M03Rik; E130320P19Rik; mKIAA4141; neurogenic extracellular slit protein; SLIL3; Slit; slit guidance ligand 2; slit homolog 2 (Drosophila); slit homolog 2 protein; Slit homolog 2 protein C-product; Slit homolog 2 protein N-product; SLIT2; Slit-2; Slit2 protein
Common Name SLIT2
Gene Symbol SLIT2
Biological Activity Determined by its ability to inhibit MC3T3/E1 osteoblasts cell differentiation.
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence QACPAQCSCS GSTVDCHGLA LRSVPRNIPR NTERLDLNGN NITRITKTDF AGLRHLRVLQ LMENKISTIE RGAFQDLKEL ERLRLNRNHL QLFPELLFLG TAKLYRLDLS ENQIQAIPRK AFRGAVDIKN LQLDYNQISC IEDGAFRALR DLEVLTLNNN NITRLSVASF NHMPKLRTFR LHSNNLYCDC HLAWLSDWLR QRPRVGLYTQ CMGPSHLRGH NVAEVQKREF VCSGHQSFMA PSCSVLHCPA ACTCSNNIVD CRGKGLTEIP TNLPETITEI RLEQNTIKVI PPGAFSPYKK LRRIDLSNNQ ISELAPDAFQ GLRSLNSLVL YGNKITELPK SLFEGLFSLQ LLLLNANKIN CLRVDAFQDL HNLNLLSLYD NKLQTIAKGT FSPLRAIQTM HLAQNPFICD CHLKWLADYL HTNPIETSGA RCTSPRRLAN KRIGQIKSKK FRCSAKEQYF IPGTEDYRSK LSGDCFADLA CPEKCRCEGT TVDCSNQKLN KIPEHIPQYT AELRLNNNEF TVLEATGIFK KLPQLRKINF SNNKITDIEE GAFEGASGVN EILLTSNRLE NVQHKMFKGL ESLKTLMLRS NRITCVGNDS FIGLSSVRLL SLYDNQITTV APGAFDTLHS LSTLNLLANP FNCNCYLAWL GEWLRKKRIV TGNPRCQKPY FLKEIPIQDV AIQDFTCDDG NDDNSCSPLS RCPTECTCLD TVVRCSNKGL KVLPKGIPRD VTELYLDGNQ FTLVPKELSN YKHLTLIDLS NNRISTLSNQ SFSNMTQLLT LILSYNRLRC IPPRTFDGLK SLRLLSLHGN DISVVPEGAF NDLSALSHLA IGANPLYCDC NMQWLSDWVK SEYKEPGIAR CAGPGEMADK LLLTTPSKKF TCQGPVDVNI LAKCNPCLSN PCKNDGTCNS DPVDFYRCTC PYGFKGQDCD VPIHACISNP CKHGGTCHLK EGEEDGFWCI CADGFEGENC EVNVDDCEDN DCENNSTCVD GINNYTCLCP PEYTGELCEE KLDFCAQDLN PCQHDSKCIL TPKGFKCDCT PGYVGEHCDI DFDDCQDNKC KNGAHCTDAV NGYTCICPEG YSGLFCEFSP PMV
Content And Storage -20°C
Activity Determined by its ability to inhibit MC3T3/E1 osteoblasts cell differentiation.
Expression System HEK293 cells
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.