missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human SLC6A14 (aa 147-233) Control Fragment Recombinant Protein

Product Code. 30196826
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30196826

Brand: Invitrogen™ RP92172

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (70%), Rat (70%). This recombinant protein control fragment may be used for blocking experiments. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes a member of the solute carrier family 6. Members of this family are sodium and chloride dependent neurotransmitter transporters. The encoded protein transports both neutral and cationic amino acids.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9UN76
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 11254
Name Human SLC6A14 (aa 147-233) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 1110007A17Rik; 9030613J17Rik; amino acid transporter ATB0+; amino acid transporter B0; amino acid transporter B0+; ATB(0+); Atb0+; ATB0plus; BMIQ11; carrier protein; CATB0plus; colonic system B0+ amino acid transporter; colonic system B0+ amino acid transporter CATB0+; LOW QUALITY PROTEIN: sodium- and chloride-dependent neutral and basic amino acid transporter B(0+); neutral cationic amino acid transporter B0,+; SLC6A14; Sodium- and chloride-dependent neutral and basic amino acid transporter B(0+); solute carrier family 6 (amino acid transporter), member 14; solute carrier family 6 (neurotransmitter transporter), member 14; solute carrier family 6 member 14
Common Name SLC6A14
Gene Symbol SLC6A14
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence FQSELPWKNCSSWSDKNCSRSPIVTHCNVSTVNKGIQEIIQMNKSWVDINNFTCINGSEIYQPGQLPSEQYWNKVALQRSSGMNETG
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.