missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human SLC4A8 (aa 41-99) Control Fragment Recombinant Protein

Product Code. 30198772
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30198772

Brand: Invitrogen™ RP109658

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (98%), Rat (98%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-139959 (PA5-139959. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Mediates electroneutral sodium- and carbonate-dependent choride-HCO3(-) exchange with a Na(+):HCO3(-) stoichiometry of 2:1. Plays a major role in pH regulation in neurons. May be involved in cell pH regulation by transporting HCO3(-) from blood to cell. Enhanced expression in severe acid stress could be important for cell survival by mediating the influx of HCO3(-) into the cells. Also mediates lithium-dependent HCO3(-) cotransport. May be regulated by osmolarity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q2Y0W8
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 9498
Name Human SLC4A8 (aa 41-99) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AW493845; C230026C11; Electroneutral Na(+)-driven Cl-HCO3 exchanger; electroneutral Na+-driven Cl-HCO3 exchanger; Electroneutral sodium bicarbonate exchanger 1; KIAA0739; k-NBC3; kNBC-3; Na-driven Cl-HCO3 exchanger NDCBE1-A; Na-driven Cl-HCO3 exchanger NDCBE1-B; Na-driven Cl-HCO3 exchanger NDCBE1-C; NBC; NBC3; NDCBE; NDCBE1; RGD:735164}; Slc4a8; slc4a8 {ECO:0000312; sodium bicarbonate cotransporter isoform 3 kNBC-3; solute carrier family 4 (anion exchanger), member 8; solute carrier family 4 member 8; solute carrier family 4, sodium bicarbonate cotransporter, member 8
Common Name SLC4A8
Gene Symbol Slc4a8
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence ELEGHRTLYVGVRMPLGRQSHRHHRTHGQKHRRRGRGKGASQGEEGLEALAHDTPSQRV
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.