missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human SLC4A4 (aa 609-685) Control Fragment Recombinant Protein

Product Code. 30211248
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30211248

Brand: Invitrogen™ RP96607

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (83%), Rat (83%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-57345 (PA5-57345. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Slc4a4 encodes a sodium bicarbonate cotransporter (NBC) involved in the regulation of bicarbonate secretion and absorption and intracellular pH. Slc4a4 is an electrogenic sodium/bicarbonate cotransporter with a Na(+):HCO3(-) stoichiometry varying from 1:2 to 1:3. Moreover, Slc4a4 may regulate bicarbonate influx/efflux at the basolateral membrane of cells and regulate intracellular pH. Isoform 2: May have a higher activity than isoform 1. Mutations in the Slc4a4 gene are associated with proximal renal tubular acidosis. Multiple transcript variants encoding different isoforms have been found for the Slc4a4 gene. Diseases associated with SLC4A4 include Renal Tubular Acidosis, Proximal, With Ocular Abnormalities and Proximal Renal Tubular Acidosis.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9Y6R1
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 8671
Name Human SLC4A4 (aa 609-685) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AI835705; Electrogenic sodium bicarbonate cotransporter 1; electrogenic sodium bicarbonate cotransporter NBCe1 variant D; hhNMC; HNBC 1; HNBC1; KNBC; kNBC 1; kNBC1; Na(+)/HCO3(-) cotransporter; Na+HCO3- cotransporter 4; NBC; NBC 1; NBC 2; NBC1; NBC2; Nbc4; NBCE 1; NBCE1; NBCe1-A; NBC-like protein; OTTHUMP00000160355; OTTHUMP00000218884; OTTHUMP00000218885; pancreas sodium bicarbonate cotransporter; PNBC; Rnbc1; SLC4A4; SLC4A5; sodium bicarbonate cotransporter; sodium bicarbonate cotransporter 1 (sodium bicarbonate cotransporter, kidney; sodium bicarbonate cotransporter, pancreas); solute carrier family 4 (anion exchanger), member 4; solute carrier family 4 (sodium bicarbonate cotransporter), member 4; solute carrier family 4 member 4; solute carrier family 4, member 4; solute carrier family 4, sodium bicarbonate cotransporter, member 4; solute carrier family 4, sodium bicarbonate cotransporter, member 4, brain type; solute carrier family 4, sodium bicarbonate cotransporter, member 5
Common Name SLC4A4
Gene Symbol SLC4A4
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence DYYPINSNFKVGYNTLFSCTCVPPDPANISISNDTTLAPEYLPTMSSTDMYHNTTFDWAFLSKKECSKYGGNLVGNN
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.