missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human SLC43A2 (aa 538-569) Control Fragment Recombinant Protein

Product Code. 30193660
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30193660

Brand: Invitrogen™ RP93382

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (88%), Rat (88%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-54451 (PA5-54451. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Uniporter that mediates the transport of the stereospecific L-phenylalanine, L-methionine and L-branched-chain amino acids, between the extracellular space and the cytoplasm and may control the transepithelial (re)absorption of neutral amino acid in kidney and small intestine. The transport activity is mediated through facilitated diffusion and is sodium ions-, chloride ions- and pH-independent.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8N370
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 124935
Name Human SLC43A2 (aa 538-569) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 7630402D21Rik; BC042513; large neutral amino acids transporter small subunit 4; Lat4; L-type amino acid transporter 4; L-type amino L-acid transporter 4; PP7664; Slc43a2; solute carrier family 43 (amino acid system L transporter), member 2; solute carrier family 43 member 2; solute carrier family 43, member 2
Common Name SLC43A2
Gene Symbol SLC43A2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence YRRQLERQLQQRQEDDKLFLKINGSSNQEAFV
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.