missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human SLC3A1 (aa 458-607) Control Fragment Recombinant Protein

Product Code. 30204190
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30204190

Brand: Invitrogen™ RP90266

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (81%), Rat (81%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-58356 (PA5-58356. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes a type II membrane glycoprotein which is one of the components of the renal amino acid transporter which transports neutral and basic amino acids in the renal tubule and intestinal tract.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q07837
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 6519
Name Human SLC3A1 (aa 458-607) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias amino acid transport related protein; amino acid transporter 1; amino acid transporter SLC3A1; ATR1; b(0,+)-type amino acid transport protein; CSNU1; D2; D2H; FLJ34681; membrane glycoprotein; NAA-TR; Nbat; Neutral and basic amino acid transport protein rBAT; NTAA; rBAT; rBAT protein; si:dkey-202m23.2; Slc3a1; SLC3A1 variant B; SLC3A1 variant C; SLC3A1 variant D; SLC3A1 variant E; SLC3A1 variant F; SLC3A1 variant G; solute carrier family 3 (amino acid transporter heavy chain), member 1; solute carrier family 3 (cystine, dibasic and neutral amino acid transporters), member 1; solute carrier family 3 (cystine, dibasic and neutral amino acid transporters, activator of cystine, dibasic and neutral amino acid transport), member 1; solute carrier family 3 member 1; solute carrier family 3, member 1
Common Name SLC3A1
Gene Symbol SLC3A1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence GNQYVNVMNMLLFTLPGTPITYYGEEIGMGNIVAANLNESYDINTLRSKSPMQWDNSSNAGFSEASNTWLPTNSDYHTVNVDVQKTQPRSALKLYQDLSLLHANELLLNRGWFCHLRNDSHYVVYTRELDGIDRIFIVVLNFGESTLLNL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.