missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human SLC39A7 (aa 30-91) Control Fragment Recombinant Protein

Product Code. 30200693
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30200693

Brand: Invitrogen™ RP101736

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (75%), Rat (75%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-62893 (PA5-62893. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The zinc transporter ZIP7, also known as SLC39A7, is a member of a family of divalent ion transporters. Zinc is an essential ion for cells and plays significant roles in the growth, development, and differentiation. ZIP7 was initially identified while characterizing genes in the major histocompatibility complex on chromosome 17. ZIP7 mRNA is abundantly and widely expressed and the protein localizes to the Golgi apparatus. It functions to transport intracellular zinc from the Golgi apparatus to the cytoplasm of the cell. ZIP7 expression is expressed by zinc. ZIP7 has been suggested to act a hub for tyrosine kinase activation and may thus be a potential therapeutic target for diseases such as cancer where prevention of tyrosine kinase activation would be advantageous.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q92504
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 7922
Name Human SLC39A7 (aa 30-91) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AA408174; AI117660; AL024048; D6S115E; D6S2244E; DADB-100D22.3; H2-K region expressed gene 4; H-2 Ke4; H2-KE4; Histidine-rich membrane protein; histidine-rich membrane protein Ke4; HKE4; HLA class II region expressed gene KE4; KE4; Ke-4; Ke4 gene; Ke4 gene, mouse, human homolog of; really interesting new gene 5 protein; Ring5; RT1 class I, locus Ke4; RT1 region expressed gene KE4; Rt1Ke4; RT1-Ke4; Slc39a7; solute carrier family 39 (zinc transporter), member 7; solute carrier family 39 member 7; Unknown (protein for MGC:142379); zinc transporter SLC39A7; ZIP7; zrt-, Irt-like protein 7
Common Name SLC39A7
Gene Symbol SLC39A7
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence GHDDLHDDLQEDFHGHSHRHSHEDFHHGHSHAHGHGHTHESIWHGHTHDHDHGHSHEDLHHG
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.