missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human SLC37A4 (aa 189-219) Control Fragment Recombinant Protein
missing translation for 'orderingAttributeHoverText'
Quantity:
100 μL
missing translation for 'unitSize'
100µL
Beschreibung
Highest antigen sequence indentity to the following orthologs: Mouse (87%), Rat (87%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-58600 (PA5-58600. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
SLC37A4 transports glucose-6-phosphate from the cytoplasm to the lumen of the endoplasmic reticulum. It forms with glucose-6-phosphatase the complex responsible for glucose production through glycogenolysis and gluconeogenesis. Hence, it plays a central role in homeostatic regulation of blood glucose levels.
Spezifikation
Spezifikation
| Accession Number | O43826 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 2542 |
| Name | Human SLC37A4 (aa 189-219) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | G6PT; G6PT1; G6PT2; G6PT3; glucose 6-phosphate transporter; glucose-5-phosphate transporter; glucose-6-phosphatase transport protein 1; glucose-6-phosphatase, transport (glucose) protein 3; glucose-6-phosphatase, transport (glucose-6-phosphate) protein 1; glucose-6-phosphatase, transport (phosphate/pyrophosphate) protein 2; glucose-6-phosphatase, transport protein 1; glucose-6-phosphate exchanger SLC37A4; Glucose-6-phosphate translocase; GSD1b; GSD-1 b; GSD1c; GSD1d; mG6PT; MGC15729; microsomal glucose-6-phosphate transporter; PRO0685; SLC37A4; solute carrier family 37 (glucose-6-phosphate transporter), member 4; solute carrier family 37 (glycerol-6-phosphate transporter), member 4; solute carrier family 37 member 4; Transformation-related gene 19 protein; transporter; TRG19; TRG-19; Unknown (protein for MGC:137413) |
| Common Name | SLC37A4 |
| Mehr anzeigen |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur