missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human SLC35D2 (aa 312-334) Control Fragment Recombinant Protein

Product Code. 30207498
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30207498 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30207498 Supplier Invitrogen™ Supplier No. RP103541
 Restricted Item
Estimated Shipment: 15-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (83%), Rat (83%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-64148. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Antiporter transporting nucleotide sugars such as UDP-N-acetylglucosamine (UDP-GlcNAc), UDP-glucose (UDP-Glc) and GDP-mannose (GDP-Man) pooled in the cytosol into the lumen of the Golgi in exchange for the corresponding nucleosides monophosphates (UMP for UDP-sugars and GMP for GDP-sugars). May take part in heparan sulfate synthesis by supplying UDP-Glc-NAc, the donor substrate, and thus be involved in growth factor signaling.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q76EJ3
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 11046
Name Human SLC35D2 (aa 312-334) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias 5730408I21Rik; fringe connection; hfrc; HFRC1; Homolog of Fringe connection protein 1; mUGTrel8; SLC35D2; solute carrier family 35 (UDP-GlcNAc/UDP-glucose transporter), member D2; solute carrier family 35 member D2; solute carrier family 35, member D2; SQV7L; SQV7-like protein; UDP-galactose transporter-related protein 8; UDP-glucuronic acid/UDP-N-acetylgalactosamine transporter; UDP-N-acetylglucosamine transporter; UDP-N-acetylglucosamine/UDP-glucose/GDP-mannose transporter; UGTrel8
Common Name SLC35D2
Gene Symbol SLC35D2
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence YSFLTLSSQLKPKPVGEENICLD
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.