missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human SLC34A3 (aa 1-78) Control Fragment Recombinant Protein

Product Code. 30207549
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30207549

Brand: Invitrogen™ RP93101

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (63%), Rat (63%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-54883 (PA5-54883. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes a member of SLC34A transporter family of proteins, and is expressed primarily in the kidney. It is involved in transporting phosphate into cells via sodium cotransport in the renal brush border membrane, and contributes to the maintenance of inorganic phosphate concentration in the kidney. Mutations in this gene are associated with hereditary hypophosphatemic rickets with hypercalciuria. Alternatively spliced transcript variants varying in the 5' UTR have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8N130
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 142680
Name Human SLC34A3 (aa 1-78) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AI649385; HHRH; Na(+)/Pi cotransporter 2 C; Na(+)-dependent phosphate cotransporter 2 C; Na+/Pi-cotransporter type IIc; naPi-2 c; Npt2c; Npt2c-v1; NptIIc; Slc34a3; Sodium/inorganic phosphate cotransporter IIC; sodium/phosphate cotransporter 2 C; Sodium-dependent phosphate transport protein 2 C; sodium-dependent phosphate transport protein 2 C; LOW QUALITY PROTEIN: sodium-dependent phosphate transport protein 2 C; sodium-phosphate cotransporter IIC; sodium-phosphate transport protein 2 C; solute carrier family 34 (sodium phosphate), member 3; solute carrier family 34 (type II sodium/phosphate contransporter), member 3; solute carrier family 34 (type II sodium/phosphate cotransporter), member 3; solute carrier family 34 member 3; type IIc Na+/Pi cotransporter; type IIc Na+/Pi-cotransporter; type IIc NaPi-cotransporter-v1
Common Name SLC34A3
Gene Symbol SLC34A3
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence MPSSLPGSQVPHPTLDAVDLVEKTLRNEGTSSSAPVLEEGDTDPWTLPQLKDTSQPWKELRVAGRLRRVAGSVLKACG
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.