missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Human SLC31A1 Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
£195.00 - £469.00
Specifications
| Antigen | SLC31A1/CTR1 |
|---|---|
| Clone | 2938A |
| Dilution | Flow Cytometry 0.25 μg/106 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30018998
|
Novus Biologicals™
MAB11484-SP |
25 μg |
£195.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30018992
|
Novus Biologicals™
MAB11484-100 |
100 μg |
£469.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SLC31A1/CTR1 Monoclonal antibody specifically detects SLC31A1/CTR1 in Human samples. It is validated for Flow CytometrySpecifications
| SLC31A1/CTR1 | |
| Flow Cytometry 0.25 μg/106 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. See Certificate of Analysis for details. *Small pack size (-SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| Copper transporter 1, COPT1, COPT1high affinity copper uptake protein 1, CTR1, CTR1MGC75487, HCTR1, hCTR1copper transport 1 homolog, solute carrier family 31 (copper transporters), member 1, Solute carrier family 31 member 1 | |
| Synthetic peptide, Accession # O15431 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. For liquid material, refer to CoA for concentration. |
| 2938A | |
| Flow Cytometry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| O15431 | |
| 1317 | |
| IgG | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze/thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title