missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human SLC28A2 (aa 590-658) Control Fragment Recombinant Protein

Product Code. 30205006
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30205006

Brand: Invitrogen™ RP103224

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (72%), Rat (72%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-63151 (PA5-63151. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The concentrative nucleoside transporter (CNT) family comprises three members: CNT1, CNT2 and CNT3. CNT2 participates in the absorption and disposition of endogenous nucleosides and mediates the first step of nucleotide biosynthesis. CNT2 levels are highly dependent on insulin (but not glucose) concentration, and the protein is under the control of the Adenosine 1 receptor. CNT family members are imperative in the response of cells to a variety of anticancer and antiviral nucleoside analogs, as the CNT proteins modulate their entry into target tissues. Increasing evidence also suggests that CNT2 may have a role in energy metabolism because activation of CNT2 relies on the opening of ATP-sensitive K+ channels.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O43868
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 9153
Name Human SLC28A2 (aa 590-658) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 2010208B10Rik; B430217P18; BB152493; CNT 2; CNT2; CNT-2; Concentrative nucleoside transporter 2; HCNT2; HsT17153; Na(+)/nucleoside cotransporter 2; purine-selective Na+ nucleoside cotransporter; rCNT2; Slc28a2; sodium/nucleoside cotransporter 2; sodium/purine nucleoside cotransporter; Sodium/purine nucleoside co-transporter; Sodium-coupled nucleoside transporter 2; solute carrier family 28 (concentrative nucleoside transporter), member 2; solute carrier family 28 (sodium-coupled nucleoside transporter), member 2; solute carrier family 28 member 2; solute carrier family 28, member 2; SPNT; SPNT1
Common Name SLC28A2
Gene Symbol SLC28A2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence GAEADCVSFPNTSFTNRTYETYMCCRGLFQSTSLNGTNPPSFSGPWEDKEFSAMALTNCCGFYNNTVCA
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.