missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human SLC28A1 (aa 1-68) Control Fragment Recombinant Protein

Product Code. 30211820
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30211820

Brand: Invitrogen™ RP90997

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (65%), Rat (65%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibodies, PA5-54023 (PA5-54023, PA5-139830 (PA5-139830. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Nucleosides play a role in signaling in several physiologic systems, and synthetic analogs of natural nucleosides are often used to treat neoplastic and viral diseases. Plasma membrane transport of nucleosides is mediated by equilibrative and concentrative nucleoside transporters, which may have specificity for purines or pyrimidines. The deduced human 650 amino acid concentrative nucleoside transporter 1 (CNT1) protein is 83% identical to the rat protein and is expressed in the intestine, kidney and liver. CNT1, also designated solute carrier family 28 (sodium-coupled nucleoside transporter), member 1 (SLC28A1), expedites sodium-dependent fluxes of uridine, azidodeoxythymidine (AZT) and adenosine, but not of guanosine or deoxyadenosine, which undergo net renal secretion. CNT1 activity may serve as a putative mechanism for renal reabsorption of physiologic nucleosides and synthetic nucleoside drugs.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O00337
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 9154
Name Human SLC28A1 (aa 1-68) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias CNT 1; CNT1; Concentrative nucleoside transporter 1; HCNT1; Na(+)/nucleoside cotransporter 1; SLC28A1; sodium/nucleoside cotransporter 1; Sodium-coupled nucleoside transporter 1; solute carrier family 28 (concentrative nucleoside transporter), member 1; solute carrier family 28 (sodium-coupled nucleoside transporter), member 1; solute carrier family 28 member 1; solute carrier family 28, member 1
Common Name SLC28A1
Gene Symbol Slc28a1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence MENDPSRRRESISLTPVAKGLENMGADFLESLEEGQLPRSDLSPAEIRSSWSEAAPKPFSRWRNLQPA
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.