missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human SLC25A20 (aa 33-73) Control Fragment Recombinant Protein

Product Code. 30207400
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30207400

Brand: Invitrogen™ RP91121

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (93%), Rat (93%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-53508 (PA5-53508. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

SLC25A20 is one of several closely related mitochondrial-membrane carrier proteins that shuttle substrates between cytosol and the intramitochondrial matrix space. It mediates the transport of acylcarnitines into mitochondrial matrix for their oxidation by the mitochondrial fatty acid-oxidation pathway. Mutations in this gene are associated with carnitine-acylcarnitine translocase deficiency, which can cause a variety of pathological conditions such as hypoglycemia, cardiac arrest, hepatomegaly, hepatic dysfunction and muscle weakness, and is usually lethal in new born and infants. This gene product is one of several closely related mitochondrial-membrane carrier proteins that shuttle substrates between cytosol and the intramitochondrial matrix space. This protein mediates the transport of acylcarnitines into mitochondrial matrix for their oxidation by the mitochondrial fatty acid-oxidation pathway. Mutations in this gene are associated with carnitine-acylcarnitine translocase deficiency, which can cause a variety of pathological conditions such as hypoglycemia, cardiac arrest, hepatomegaly, hepatic dysfunction and muscle weakness, and is usually lethal in new born and infants.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O43772
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 788
Name Human SLC25A20 (aa 33-73) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 1110007P09Rik; C78826; CAC; CACT; carnitine/acylcarnitine carrier protein; carnitine/acylcarnitine translocase; mCAC; mitochondrial carnitine/acylcarnitine carrier protein; mitochondrial carnitine-acylcarnitine translocase; slc25a20; solute carrier family 25 (carnitine/acylcarnitine translocase), member 20; solute carrier family 25 (mitochondrial carnitine/acylcarnitine translocase), member 20; solute carrier family 25 member 20; Unknown (protein for MGC:128410); zgc:77760
Common Name SLC25A20
Gene Symbol SLC25A20
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence TVKVRLQTQPPSLPGQPPMYSGTFDCFRKTLFREGITGLYR
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.