missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human SLC22A2/OCT2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB6547
This item is not returnable.
View return policy
Description
SLC22A2/OCT2 Monoclonal specifically detects SLC22A2/OCT2 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry.
Specifications
| SLC22A2/OCT2 | |
| Monoclonal | |
| LYOPH | |
| Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 8-25 ug/mL | |
| O15244 | |
| SLC22A2 | |
| NS0 mouse myeloma cell line transfected with human SLC-22A2 Accession # O15244 | |
| 100 μg | |
| Primary | |
| Detects human SLC22A2 in direct ELISAs. | |
| Human | |
| Purified |
| Flow Cytometry, Immunohistochemistry | |
| 640438 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| hOCT2, MGC32628, OCT2, OCT2Organic cation transporter 2, solute carrier family 22 (organic cation transporter), member 2, solute carrier family 22 member 2 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6582 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction