missing translation for 'onlineSavingsMsg'
Learn More

Human SLC1A5 Antibody, R&D Systems™

Rabbit Monoclonal Antibody

£206.00 - £479.00

Specifications

Antigen SLC1A5
Clone 3085B
Dilution Immunohistochemistry 1-10 μg/mL, Immunocytochemistry 1-10 μg/mL
Applications Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30279910
View Documents
R&D Systems
MAB11628-SP
25 μg
£206.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30279895
View Documents
R&D Systems
MAB11628-100
100 μg
£479.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

SLC1A5 Monoclonal antibody specifically detects SLC1A5 in Human samples. It is validated for Immunohistochemistry,Immunocytochemistry
Specifications

Specifications

SLC1A5
Immunohistochemistry 1-10 μg/mL, Immunocytochemistry 1-10 μg/mL
Monoclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
AAAT, Alanine/serine/cysteine transporter 2, ASC amino acid transporter-2, ASCT2, ATB(0), ATBO, baboon M7 virus receptor, M7V1, M7VS1, neutral amino acid transporter B, neutral amino acid transporter B(0), R16, RD114 virus receptor, RD114/simian type D retrovirus receptor, RDR, RDRC, Sodium-dependent neutral amino acid transporter type 2, solute carrier family 1 (neutral amino acid transporter), member 5, Solute carrier family 1 member 5
Synthetic Peptide, Accession # Q9JKD8
Primary
Reconstitute lyophilized material at 0.2mg/mL in sterile PBS. For liquid material, refer to CoA for concentration.
3085B
Immunohistochemistry, Immunocytochemistry
Unconjugated
Rabbit
Human
Q9JKD8
6510
IgG
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20°C to -70 °C under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.