missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human SLC14A1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£387.00
Specifications
| Antigen | SLC14A1 |
|---|---|
| Clone | 888418 |
| Concentration | LYOPH |
| Dilution | Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready |
| Applications | Flow Cytometry, CyTOF |
Description
SLC14A1 Monoclonal specifically detects SLC14A1 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
| SLC14A1 | |
| LYOPH | |
| Flow Cytometry, CyTOF | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q13336 | |
| 6563 | |
| NS0 mouse myeloma cell line transfected with human SLC14A1 Accession # Q13336 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human SLC14A1 in direct ELISAs. |
| 888418 | |
| Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| FLJ33745, HsT1341, HUT11, JK, JKFLJ41687, RACH1, RACH2, solute carrier family 14 (urea transporter), member 1 (Kidd blood group), Solute carrier family 14 member 1, truncated urea transporter, urea transporter 1, urea transporter JK glycoprotein, Urea transporter, erythrocyte, urea transporter-B1, UT1, UT1UT-B1, UT-B1, UTE, UTEblood group Kidd urea transporter | |
| SLC14A1 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title