missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human SLAM/CD150 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB164
This item is not returnable.
View return policy
Description
SLAM/CD150 Monoclonal specifically detects SLAM/CD150 in Human samples. It is validated for Western Blot.
Specifications
| SLAM/CD150 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q13291 | |
| SLAMF1 | |
| Mouse myeloma cell line NS0-derived recombinant human SLAM/CD150 Ala21-Pro237 (predicted) Accession # Q13291 | |
| 500 μg | |
| Primary | |
| Detects human SLAM/CD150 in direct ELISA and Western blots. In direct ELISAs, no cross-reactivity with recombinant human 2B4 is observed. | |
| Human | |
| Purified |
| Western Blot | |
| 123317 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD 150, CD150, CD150IPO-3, CDw150, IPO-3, signaling lymphocytic activation molecule, signaling lymphocytic activation molecule family member 1, SLAMCD150 antigen, SLAMF1 | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 6504 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction