missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Sirtuin 1/SIRT1 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF7714
This item is not returnable.
View return policy
Description
Sirtuin 1/SIRT1 Polyclonal specifically detects Sirtuin 1/SIRT1 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready, Knockout Validated.
Specifications
| Sirtuin 1/SIRT1 | |
| Polyclonal | |
| Unconjugated | |
| Q96EB6 | |
| SIRT1 | |
| E. coli-derived recombinant human Sirtuin 1/SIRT1 Ala2-Ser747 Accession # Q96EB6 | |
| 100 μg | |
| Primary | |
| Detects human Sirtuin 1/SIRT1 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human SIRT2 is observed. | |
| Human | |
| IgG |
| Western Blot, Flow Cytometry, CyTOF, KnockDown | |
| LYOPH | |
| Western Blot 0.2 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready, Knockout Validated | |
| EC 3.5.1, S. cerevisiae, homolog) 1, SIR2alpha, SIR2L1, SIR2-like protein 1, SIRT1, sirtuin 1 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 23411 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu