missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human SIRP beta 2 Alexa Fluor 350-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB9998U-100UG
This item is not returnable.
View return policy
Description
SIRPB2 Monoclonal specifically detects SIRPB2 in Human samples. It is validated for Flow Cytometry.
Specifications
| SIRPB2 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| dJ776F14.2, Protein Tyrosine Phosphatase Non-Receptor Type Substrate 1-Like 3, Protein Tyrosine Phosphatase Non-Receptor Type Substrate Protein, Protein Tyrosine Phosphatase, Non-Receptor Type 1-Like, Protein Tyrosine Phosphatase, Non-Receptor Type Substrate 1-Like 3, PTPN1L, PTPNS1L3, Signal-Regulatory Protein Beta 2, Signal-Regulatory Protein Beta-2, SIRP-beta-2, SIRPG | |
| HEK293 human embryonic kidney cell line, HEK293 derived human SIRP beta 2, Gln33-Gly287, Accession # Q5JXA9 | |
| 100 μg | |
| 284759 | |
| Human | |
| Supernatant |
| Flow Cytometry | |
| 1008709 | |
| Flow Cytometry 0.25-1 μg/106 cells | |
| Q5JXA9 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human SIRP beta 2 in direct ELISAs. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction