missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human SIRP beta 1/gamma Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB2096-SP
This item is not returnable.
View return policy
Description
SIRP beta 1/gamma Monoclonal specifically detects SIRP beta 1/gamma in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
Specifications
| SIRP beta 1/gamma | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| O00241.4 | |
| SIRPB1 | |
| Mouse myeloma cell line NS0-derived recombinant human SIRP beta 1, Gly26-Ala369 (Arg53His and Ala363Pro), Accession # O00241.4 | |
| 25 μg | |
| Primary | |
| Detects human SIRP beta 1 and human SIRP gamma in direct ELISAs and Western blots. | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, CyTOF | |
| 308906 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD172 antigen-like family member B, CD172b, CD172b antigen, DKFZp686A05192, FLJ26614, signal-regulatory protein beta 1, signal-regulatory protein beta-1, SIRPB1, SIRP-BETA-1, SIRP-beta-1 isoform 3 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 10326 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction