missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human SIRP beta 1/CD172b Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB20961-100
This item is not returnable.
View return policy
Description
SIRP beta 1/CD172b Monoclonal specifically detects SIRP beta 1/CD172b in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF-ready.
Specifications
| SIRP beta 1/CD172b | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS. | |
| CD172 antigen-like family member B, CD172b, CD172b antigen, DKFZp686A05192, FLJ26614, signal-regulatory protein beta 1, signal-regulatory protein beta-1, SIRPB1, SIRP-BETA-1, SIRP-beta-1 isoform 3 | |
| Mouse myeloma cell line NS0-derived recombinant human SIRP beta 1/CD172b, Gly26-Ala369 (Arg53His and Ala363Pro), Accession # O00241.4 | |
| 100 μg | |
| Primary | |
| Detects human SIRP beta 1/CD172b in direct ELISAs. | |
| Human | |
| Purified |
| Flow Cytometry, Immunocytochemistry, CyTOF | |
| 308908 | |
| Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL, CyTOF-ready | |
| O00241.4 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 10326 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction