missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human IL-2R beta (soluble) Fc Recombinant Protein, PeproTech®

Product Code. 17914192
Click to view available options
Quantity:
1 mg
100 μg
25 μg
250 μg
5 μg
500 μg
Unit Size:
100µg
1mg
250µg
25µg
500µg
5µg
This item is not returnable. View return policy

Product Code. 17914192

Brand: Gibco™ 20002RB1MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Recombinant Human sIL-2 Receptor beta Fc is a 53.5 kDa protein of 473 amino acids, consisting of the extracellular domain of the mature sequence, as well as a 230 C-terminal Fc region linked by two glycines. As a result of glycosylation, the Recombinant Human sIL-2 Receptor beta Fc migrates to an approximate molecular weight of 60-65 kDa, under reducing conditions. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

CD122 (Interleukin 2 receptor) is involved in T cell-mediated immune responses, primarily expressed in the hematopoietic system and is present in 3 forms. The low affinity form of CD122 is a monomer of the alpha subunit and is not involved in signal transduction. The intermediate affinity form consists of an alpha/beta subunit heterodimer, while the high affinity form consists of an alpha/beta/gamma subunit heterotrimer. Both the intermediate and high affinity forms of the receptor are involved in receptor-mediated endocytosis and transduction of mitogenic signals from CD122. The protein encoded by the CD122 gene represents the beta subunit and is a type I membrane protein. Diseases associate with CD122 protein dysfunction include oligoarticular juvenile idiopathic arthritis and rheumatoid factor negative juvenile idiopathic arthritis.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P14784
For Use With (Application) Functional Assay
Format Lyophilized
Formulation protein with no preservative
Gene ID (Entrez) 3560
Molecular Weight (g/mol) 50.8 kDa
Name Human IL-2 R beta (soluble) Fc
Quantity 1 mg
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias CD122; CD122 antigen; high affinity IL-2 receptor beta subunit; high affinity IL-2 receptor subunit beta; IL-15 receptor beta chain; IL15RB; IL15Rbeta; IL-15 Rbeta; IL-2 receptor beta chain; IL-2 receptor subunit beta; IL-2/15 receptor-beta; Il-2/15 Rbeta; IL-2 R subunit beta; IL2RB; IL-2 RB; IL2RBC; Il-2 Rbeta; interleukin 15 receptor, beta; interleukin 2 receptor subunit beta; interleukin 2 receptor, beta; interleukin 2 receptor, beta chain; Interleukin-15 receptor subunit beta; Interleukin-2 receptor subunit beta; p70; p70-75; p75
Common Name CD122
Biological Activity Determined by its ability to inhibit human IL-15 induced cell proliferation in MO7e cells. In the presence of 4 ng/ml of recombinant human IL-15, the expected ED50 for this effect is between 1.0-4.0 ug/ml
Conjugate Unconjugated
Recombinant Recombinant
Protein Tag Fc-tag
Sequence AVNGTSQFTC FYNSRANISC VWSQDGALQD TSCQVHAWPD RRRWNQTCEL LPVSQASWAC NLILGAPDSQ KLTTVDIVTL RVLCREGVRW RVMAIQDFKP FENLRLMAPI SLQVVHVETH RCNISWEISQ ASHYFERHLE FEARTLSPGH TWEEAPLLTL KQKQEWICLE TLTPDTQYEF QVRVKPLQGE FTTWSPWSQP LAFRTKPAAL GKDTGGPKSC DKTHTCPPCP APELLGGPSV FLFPPKPKDT LMISRTPEVT CVVVDVSHED PEVKFNWYVD GVEVHNAKTK PREEQYNSTY RVVSVLTVLH QDWLNGKEYK CKVSNKALPA PIEKTISKAK GQPREPQVYT LPPSRDELTK NQVSLTCLVK GFYPSDIAVE WESNGQPENN YKTTPPVLDS DGSFFLYSKL TVDKSRWQQG NVFSCSVMHE ALHNHYTQKS LSLSPGK
Content And Storage -20°C
Expression System CHO cells
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.