missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human IL-12 Receptor beta-2 (soluble) Fc Recombinant Protein, PeproTech®

Product Code. 17964322
Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
500 μg
Unit Size:
100µg
1mg
20µg
250µg
500µg
This item is not returnable. View return policy

Product Code. 17964322

Brand: Gibco™ 20012RB220UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Recombinant Human sIL-12 Receptor beta-2 Fc has a calculated molecular weight of 94.9 kDa, and contains 839 amino acid residues, which consists of the extracellular domain of IL-12R beta-2 fused to the Fc portion of human IgG and contains a C-terminal His-tag. Due to glycosylation, it migrates at an approximate molecular weight of 116 kDa, under reducing conditions. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

The protein encoded by this gene is a type I transmembrane protein identified as a subunit of the interleukin 12 receptor complex. The coexpression of this and IL12RB1 proteins was shown to lead to the formation of high-affinity IL12 binding sites and reconstitution of IL12 dependent signaling. The expression of this gene is up-regulated by interferon gamma in Th1 cells, and plays a role in Th1 cell differentiation. The up-regulation of this gene is found to be associated with a number of infectious diseases, such as Crohn's disease and leprosy, which is thought to contribute to the inflammatory response and host defense.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q99665
For Use With (Application) Functional Assay
Format Lyophilized
Formulation protein with no preservative
Gene ID (Entrez) 3595
Molecular Weight (g/mol) 94.9 kDa
Name Human IL-12 Receptor beta-2 (soluble) Fc
Quantity 20 μg
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias A930027I18Rik; Ifnm; IL-12 receptor subunit beta-2; IL-12 R subunit beta-2; Il12rb2; IL-12 RB2; IL-12 R-beta-2; interferon response to microorganisms; interleukin 12 receptor subunit beta 2; interleukin 12 receptor, beta 2; interleukin-12 receptor beta-2 chain; interleukin-12 receptor subunit beta-2; RP11-102M16.1
Common Name IL12RB2
Biological Activity Human sIL-2 Receptor beta-2 Fc activity is measured by its ability to bind to Human IL-12p70 in a functional ELISA.
Conjugate Unconjugated
Recombinant Recombinant
Protein Tag Fc-tag, His-tag
Sequence CKRGDVTVKP SHVILLGSTV NITCSLKPRQ GCFHYSRRNK LILYKFDRRI NFHHGHSLNS QVTGLPLGTT LFVCKLACIN SDEIQICGAE IFVGVAPEQP QNLSCIQKGE QGTVACTWER GRDTHLYTEY TLQLSGPKNL TWQKQCKDIY CDYLDFGINL TPESPESNFT AKVTAVNSLG SSSSLPSTFT FLDIVRPLPP WDIRIKFQKA SVSRCTLYWR DEGLVLLNRL RYRPSNSRLW NMVNVTKAKG RHDLLDLKPF TEYEFQISSK LHLYKGSWSD WSESLRAQTP EEEPTGMLDV WYMKRHIDYS RQQISLFWKN LSVSEARGKI LHYQVTLQEL TGGKAMTQNI TGHTSWTTVI PRTGNWAVAV SAANSKGSSL PTRINIMNLC EAGLLAPRQV SANSEGMDNI LVTWQPPRKD PSAVQEYVVE WRELHPGGDT QVPLNWLRSR PYNVSALISE NIKSYICYEI RVYALSGDQG GCSSILGNSK HKAPLSGPHI NAITEEKGSI LISWNSIPVQ EQMGCLLHYR IYWKERDSNS QPQLCEIPYR VSQNSHPINS LQPRVTYVLW MTALTAAGES SHGNEREFCL QGKANGGPKS CDKTHTCPPC PAPELLGGPS VFLFPPKPKD TLMISRTPEV TCVVVDVSHE DPEVKFNWYV DGVEVHNAKT KPREEQYNST YRVVSVLTVL HQDWLNGKEY KCKVSNKALP APIEKTISKA KGQPREPQVY TLPPSRDELT KNQVSLTCLV KGFYPSDIAV EWESNGQPEN NYKTTPPVLD SDGSFFLYSK LTVDKSRWQQ GNVFSCSVMH EALHNHYTQK SLSLSPGKHH HHHHHH
Content And Storage -20°C
Expression System CHO cells
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.