missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Sigma-1 R Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems MAB1076-SP
This item is not returnable.
View return policy
Description
Sigma-1 R/OPRS1 Monoclonal specifically detects Sigma-1 R/OPRS1 in Human samples. It is validated for Immunocytochemistry.
Specifications
| Sigma-1 R/OPRS1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Aging-associated gene 8 protein, ALS16, FLJ25585, hSigmaR1, OPRS1, SIG-1R, sigma 1, Sigma 1-type opioid receptor, sigma non-opioid intracellular receptor 1, sigma1 receptor, sigma1R, Sigma-1R, sigma1-receptor, SIGMAR1, SR31747 binding protein 1, SR31747-binding protein, SRBP, SR-BP, SR-BP1, SRBPMGC3851, type I sigma receptor | |
| Synthetic peptide containing human Sigma-1 R, Accession # Q99720 | |
| 25 μg | |
| 10280 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG |
| Immunocytochemistry | |
| 1319A | |
| Immunocytochemistry 3-25 μg/mL | |
| Q99720 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| Primary | |
| Detects human Sigma-1 R in direct ELISAs. | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction