missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Siglec-6/CD327 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB28592-SP
This item is not returnable.
View return policy
Description
Siglec-6/CD327 Monoclonal specifically detects Siglec-6/CD327 in Human samples. It is validated for Immunohistochemistry.
Specifications
| Siglec-6/CD327 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| CD327, CD327 antigen, CD33 antigen-like 1, CD33L1, CD33L1OBBP1CD327, CD33L2, CD33LCDW327, CDw327, OB-BP1, Obesity-binding protein 1, sialic acid binding Ig-like lectin 6, sialic acid-binding Ig-like lectin 6, Siglec6, Siglec-6 | |
| Chinese Hamster Ovary cell line, CHO-derived human Siglec-6, Gln27-Ser331, Accession # O43699 | |
| 25 μg | |
| 946 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG2a |
| Immunohistochemistry | |
| 1004337 | |
| Immunohistochemistry 5-25 μg/mL | |
| O43699 | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| Primary | |
| Detects human Sigles-6 in direct ELISAs. | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction