missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Siglec-6/CD327 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£205.00 - £406.00
Specifications
| Antigen | Siglec-6/CD327 |
|---|---|
| Clone | 767335R |
| Dilution | ELISA |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18129280
|
R&D Systems
MAB28591-SP |
25 μg |
£205.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18199430
![]() |
R&D Systems
MAB28591-100 |
100 μg |
£406.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Siglec-6/CD327 Monoclonal specifically detects Siglec-6/CD327 in Human samples. It is validated for ELISA.Specifications
| Siglec-6/CD327 | |
| ELISA | |
| Monoclonal | |
| Supernatant | |
| Human | |
| O43699 | |
| 946 | |
| IgG1 | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 767335R | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Lyophilized from a 0.2μm filtered solution in PBS with Trehalose *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
| CD327, CD327 antigen, CD33 antigen-like 1, CD33L1, CD33L1OBBP1CD327, CD33L2, CD33LCDW327, CDw327, OB-BP1, Obesity-binding protein 1, sialic acid binding Ig-like lectin 6, sialic acid-binding Ig-like lectin 6, Siglec6, Siglec-6 | |
| Chinese hamster ovary cell line CHO-derived recombinant human Siglec-6/CD327 Gln27-Ser331 Accession # O43699 | |
| Primary | |
| Reconstitute at 0.5mg/mL in sterile PBS. | |
| Detects human Siglec-6/CD327 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title