missing translation for 'onlineSavingsMsg'
Learn More

Human Siglec-5/Siglec-14 Alexa Fluor™ 750-conjugated Antibody, R&D Systems™

Product Code. 18724877
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18724877 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18724877 Supplier R&D Systems Supplier No. FAB10721S100UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Siglec-5/Siglec-14 Monoclonal specifically detects Siglec-5/Siglec-14 in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Siglec-5/Siglec-14
Applications Flow Cytometry
Classification Monoclonal
Clone 194128
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 750
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. O15389
Gene Symbols SIGLEC5
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Siglec-5/Siglec-14 Lys18-Thr434 Accession # O15389
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8778
Test Specificity Detects human Siglec-5/Siglec-14 in ELISAs. In sandwich immunoassays, 100% cross-reactivity with recombinant human (rh) Siglec-14 is observed and no cross-reactivity with rhSiglec-3, rhSiglec-7, or rhSiglec-9 is observed.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.