missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Siglec-2/CD22 LlaMABodyTM Bivalent VHH HuIgG2 Fusion Antibody, R&D Systems™

Llama Monoclonal Antibody
Brand: R&D Systems LMAB107321-100
This item is not returnable.
View return policy
Description
Siglec-2/CD22 Monoclonal antibody specifically detects Siglec-2/CD22 in Human samples. It is validated for Immunocytochemistry
Specifications
| Siglec-2/CD22 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| B-cell receptor CD22, BL-CAM, B-lymphocyte cell adhesion molecule, CD22, CD22 antigenMGC130020, CD22 molecule, sialic acid binding Ig-like lectin 2, Sialic acid-binding Ig-like lectin 2, Siglec2, SIGLEC-2, SIGLEC2FLJ22814, T-cell surface antigen Leu-14 | |
| Chinese Hamster Ovary cell line CHO-derived human Siglec-2/CD22 as immunogen for bivalent Llama VHH-Human IgG2 Fusion Antibody, Asp20-Arg687, Accession # CAA42006.1 | |
| 100 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
| Immunocytochemistry | |
| L007.2.5H | |
| Immunocytochemistry 8-25 ug/mL | |
| CAA42006.1 | |
| Llama | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 933 | |
| Human | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion