missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Siglec-2/CD22 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB19681
This item is not returnable.
View return policy
Description
Siglec-2/CD22 Monoclonal specifically detects Siglec-2/CD22 in Human samples. It is validated for Western Blot.
Specifications
| Siglec-2/CD22 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.2 ug/mL | |
| CAA42006 | |
| CD22 | |
| Mouse myeloma cell line NS0-derived recombinant human Siglec-2/CD22 Asp20-Arg687 Accession # CAA42006 | |
| 100 μg | |
| Primary | |
| Detects human Siglec-2/CD22 in direct ELISAs and Western blots. In Western blots, approximately 25% cross-reactivity with recombinant human (rh) Siglec-7 and rhSiglec-9 is observed, and no cross-reactivity with rhSiglec-3 is observed. | |
| Human | |
| Purified |
| Western Blot | |
| 219902 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| B-cell receptor CD22, BL-CAM, B-lymphocyte cell adhesion molecule, CD22, CD22 antigenMGC130020, CD22 molecule, sialic acid binding Ig-like lectin 2, Sialic acid-binding Ig-like lectin 2, Siglec2, SIGLEC-2, SIGLEC2FLJ22814, T-cell surface antigen Leu-14 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 933 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction